Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001172.hg.1 |
TC6_cox_hap2000076.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001172.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000987.hg.1 |
TC6_mcf_hap5000062.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000987.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001174.hg.1 |
TC6_cox_hap2000076.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001174.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001106.hg.1 |
TC6_mann_hap4000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001106.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001155.hg.1 |
TC6_qbl_hap6000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001155.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003831.hg.1 |
TC06000382.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003831.hg.1 |
ENST00000375810.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003831.hg.1 |
ENST00000480039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003831.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000989.hg.1 |
TC6_mcf_hap5000062.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000989.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003833.hg.1 |
TC06000382.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003833.hg.1 |
ENST00000375810.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003833.hg.1 |
ENST00000480039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003833.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001117.hg.1 |
TC6_dbb_hap3000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001117.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001115.hg.1 |
TC6_dbb_hap3000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001115.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000962.hg.1 |
TC6_ssto_hap7000063.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000962.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000964.hg.1 |
TC6_ssto_hap7000063.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000964.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001153.hg.1 |
TC6_qbl_hap6000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001153.hg.1 |
NM_138277.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001108.hg.1 |
TC6_mann_hap4000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001108.hg.1 |
NM_138277.2 |
Refseq |