Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043014.hg.1 |
TC02002674.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043014.hg.1 |
ENST00000332624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043014.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043014.hg.1 |
ENST00000542292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043014.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000440597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000440597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000440597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000451703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000451703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000451703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000451703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000486291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000486291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000486291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213175.hg.1 |
TC0200015425.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000332624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000430254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000440597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000440597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000440597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000451703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000451703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000451703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000451703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000486291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000486291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000486291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
ENST00000620184.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015425.hg.1 |
NM_015049.2 |
Refseq |