Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142326.1 | NP_001135798.1 | 3 - 4 | 445 | 129 | |
NM_001142327.1 | NP_001135799.1 | 4 - 5 | 562 | 129 | ||
NM_021145.3 | NP_066968.3 | 6 - 7 | 799 | 129 | ||
NR_024549.1 | 4 - 5 | 562 | 129 | |||
NR_024550.1 | 4 - 5 | 562 | 129 | |||
XM_011516735.2 | XP_011515037.1 | 4 - 5 | 1484 | 129 | ||
XM_011516737.1 | XP_011515039.1 | 5 - 6 | 609 | 129 | ||
XM_017012865.1 | XP_016868354.1 | 5 - 6 | 1578 | 129 | ||
XM_017012868.1 | XP_016868357.1 | 5 - 6 | 609 | 129 | ||
XM_017012871.1 | XP_016868360.1 | 5 - 6 | 1578 | 129 | ||
XM_017012872.1 | XP_016868361.1 | 4 - 5 | 1484 | 129 | ||
XM_017012873.1 | XP_016868362.1 | 5 - 6 | 1373 | 129 | ||
XM_017012874.1 | XP_016868363.1 | 5 - 6 | 609 | 129 | ||
XM_017012875.1 | XP_016868364.1 | 5 - 6 | 1373 | 129 | ||
XM_017012876.1 | XP_016868365.1 | 4 - 5 | 471 | 129 | ||
XM_017012877.1 | XP_016868366.1 | 4 - 5 | 471 | 129 | ||
XM_017012878.1 | XP_016868367.1 | 6 - 7 | 1467 | 129 | ||
GenBank mRNA | AF084530.1 | - | 4 - 5 | 511 | 129 | |
AF202144.1 | - | 4 - 5 | 511 | 129 | ||
AF202145.1 | - | 4 - 5 | 511 | 129 | ||
AK126664.1 | - | 4 - 5 | 1057 | 129 | ||
AK296211.1 | - | 3 - 4 | 338 | 129 | ||
AK314622.1 | - | 4 - 5 | 498 | 129 | ||
BC029370.1 | - | 3 - 4 | 502 | 129 | ||
BC070064.1 | - | 6 - 7 | 745 | 4373991 | 129 | |
EL736930.1 | - | 2 - 3 | 236 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |