Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318785.1 | NP_001305714.1 | 13 - 14 | 1472 | 106 | |
NM_020320.4 | NP_064716.2 | 14 - 15 | 1307 | 106 | ||
NR_134857.1 | 14 - 15 | 1303 | 106 | |||
XM_011535948.2 | XP_011534250.1 | 14 - 15 | 1292 | 106 | ||
XM_011535949.2 | XP_011534251.1 | 14 - 15 | 1292 | 106 | ||
XM_017011072.1 | XP_016866561.1 | 14 - 15 | 1473 | 106 | ||
XM_017011073.1 | XP_016866562.1 | 15 - 16 | 1691 | 106 | ||
XM_017011074.1 | XP_016866563.1 | 14 - 15 | 1294 | 106 | ||
XM_017011075.1 | XP_016866564.1 | 13 - 14 | 1417 | 106 | ||
XM_017011076.1 | XP_016866565.1 | 15 - 16 | 1635 | 106 | ||
XM_017011077.1 | XP_016866566.1 | 13 - 14 | 1194 | 106 | ||
XM_017011078.1 | XP_016866567.1 | 13 - 14 | 1238 | 106 | ||
XM_017011079.1 | XP_016866568.1 | 14 - 15 | 1473 | 106 | ||
XM_017011080.1 | XP_016866569.1 | 15 - 16 | 1635 | 106 | ||
XM_017011081.1 | XP_016866570.1 | 13 - 14 | 1238 | 106 | ||
GenBank mRNA | AK023550.1 | - | 14 - 15 | 1250 | 106 | |
AK093934.1 | - | 14 - 15 | 1680 | 106 | ||
AK315669.1 | - | 14 - 15 | 1244 | 106 | ||
BC010420.1 | - | 14 - 15 | 1278 | 3613406 | 106 | |
BC105784.1 | - | 13 - 14 | 1411 | 106 | ||
CA415020.1 | - | 6 - 7 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |