Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKHD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177531.4 | NP_803875.2 | 46 - 47 | 7106 | 100 | |
XM_011517371.2 | XP_011515673.1 | 46 - 47 | 7057 | 100 | ||
XM_017013969.1 | XP_016869458.1 | 46 - 47 | 7057 | 100 | ||
XM_017013970.1 | XP_016869459.1 | 45 - 46 | 6943 | 100 | ||
XM_017013971.1 | XP_016869460.1 | 46 - 47 | 6895 | 100 | ||
XM_017013972.1 | XP_016869461.1 | 44 - 45 | 6754 | 100 | ||
XM_017013973.1 | XP_016869462.1 | 22 - 23 | 4113 | 100 | ||
XM_017013974.1 | XP_016869463.1 | 17 - 18 | 3613 | 100 | ||
GenBank mRNA | AY219181.1 | - | 46 - 47 | 7106 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |