Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047259.hg.1 |
TC01003034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047259.hg.1 |
ENST00000256649.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047259.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047259.hg.1 |
ENST00000369464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047259.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047259.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047260.hg.1 |
TC01003034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047260.hg.1 |
ENST00000256649.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047260.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047260.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047260.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215156.hg.1 |
TC0100015415.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000369464.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
ENST00000497970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_001145635.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015415.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
ENST00000256649.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
ENST00000369461.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
ENST00000485032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
NM_025188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100215157.hg.1 |
TC0100015415.hg.1 |
Clariom D TC |