Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021374.hg.1 |
TC01001363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021374.hg.1 |
ENST00000368078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021374.hg.1 |
ENST00000368079.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021374.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021375.hg.1 |
TC01001363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021375.hg.1 |
ENST00000368078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021375.hg.1 |
ENST00000368079.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021375.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021379.hg.1 |
TC01001363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021379.hg.1 |
ENST00000368078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021379.hg.1 |
ENST00000368079.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021379.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021373.hg.1 |
TC01001363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021373.hg.1 |
ENST00000368079.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021373.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021376.hg.1 |
TC01001363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021376.hg.1 |
ENST00000368078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021376.hg.1 |
ENST00000368079.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021376.hg.1 |
ENST00000441151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021376.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021377.hg.1 |
TC01001363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021377.hg.1 |
ENST00000368078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021377.hg.1 |
ENST00000368079.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021377.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175479.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175479.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175479.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175479.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175479.hg.1 |
TC0100010283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100175482.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175482.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175482.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175482.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175482.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175482.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175482.hg.1 |
TC0100010283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100175478.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175478.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175478.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175478.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175478.hg.1 |
TC0100010283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100175477.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175477.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175477.hg.1 |
TC0100010283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100175480.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175480.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175480.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100175480.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100175480.hg.1 |
TC0100010283.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000368078.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000467691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
ENST00000481081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010283.hg.1 |
NM_001231.4 |
Refseq |