Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007980.hg.1 |
TC07000547.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007980.hg.1 |
ENST00000265742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007980.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000442183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000442183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000442183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000442183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000465883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000465883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000465883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000486698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
ENST00000486698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158083.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158083.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158083.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158083.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158083.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158083.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158083.hg.1 |
TC0700008321.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000265742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000413588.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000422095.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000439883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000442183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000442183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000442183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000442183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000465883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000465883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000465883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000486698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
ENST00000486698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008321.hg.1 |
NM_019004.1 |
Refseq |