Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004045.hg.1 |
TC14002297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004045.hg.1 |
ENST00000261245.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004045.hg.1 |
ENST00000539616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004045.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004045.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000553354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000553354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000553354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000555545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000555545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000555545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000553354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000553354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000553354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000555545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000555545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000555545.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556525.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000556764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010615.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000261245.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000539616.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000554002.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000554641.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
ENST00000557134.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
NM_001177963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
NM_002431.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179462.hg.1 |
TC1400010615.hg.1 |
Clariom D TC |