Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000309295.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000526990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000527214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000527214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000527214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000527214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000527214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000527214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000527214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529056.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529099.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000529596.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000531106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000531106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000531106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533364.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
ENST00000533465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013044.hg.1 |
NM_001099409.2 |
Refseq |