Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371843.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000371847.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000396199.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000463938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000463938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000463938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000463938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000463938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000463938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000463938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000463938.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000465990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000465990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000465990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000465990.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000471975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000471975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000471975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000471975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000471975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000471975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000471975.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487543.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487543.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487543.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487543.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487543.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487543.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487543.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000487629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000490850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
ENST00000603831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286238.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_001286239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014127.hg.1 |
NM_019073.3 |
Refseq |