Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026020.hg.1 |
TC07001683.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026020.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026020.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026020.hg.1 |
ENST00000448792.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026020.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026023.hg.1 |
TC07001683.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026023.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026023.hg.1 |
ENST00000448792.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07026023.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186489.hg.1 |
TC0700012036.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000445482.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012036.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186492.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186492.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186492.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186492.hg.1 |
ENST00000249330.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186492.hg.1 |
ENST00000611537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700186492.hg.1 |
NM_003378.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700186492.hg.1 |
TC0700012036.hg.1 |
Clariom D TC |