Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019793.hg.1 |
ENST00000339364.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019793.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019793.hg.1 |
ENST00000371110.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019793.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019793.hg.1 |
TC10001559.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000468783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
ENST00000489982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011524.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
ENST00000339364.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
ENST00000371109.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
ENST00000371110.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
ENST00000467625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
NM_152309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000178354.hg.1 |
TC1000011524.hg.1 |
Clariom D TC |