Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013374.hg.1 |
TC08001051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013374.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013374.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013376.hg.1 |
TC08001051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013376.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013376.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013375.hg.1 |
TC08001051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013375.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013375.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013377.hg.1 |
TC08001051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013377.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013377.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167029.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167029.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167029.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167029.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167029.hg.1 |
TC0800009872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800167030.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167030.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167030.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167030.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167030.hg.1 |
TC0800009872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009872.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167027.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167027.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167027.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167027.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167027.hg.1 |
TC0800009872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800167028.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167028.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167028.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167028.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167028.hg.1 |
TC0800009872.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800167031.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167031.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167031.hg.1 |
ENST00000312584.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167031.hg.1 |
NM_003840.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167031.hg.1 |
TC0800009872.hg.1 |
Clariom D TC |