Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000358130.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000373250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000373250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000373250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000373250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000373250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000373250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000373250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000373250.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000430960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000436386.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000447319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000451455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000451455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000451455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000451455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000451455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000451455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000451455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000451455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000491275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000491275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000491275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000491275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000491275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000491275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000491275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
ENST00000491275.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010172.hg.1 |
NM_030763.2 |
Refseq |