Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000299275.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000317589.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000355397.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000424268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000429027.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000538714.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000539256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000542828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
ENST00000543806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
NM_001143821.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
NM_019012.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002697.hg.1 |
TC12000219.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000299275.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000424268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000429027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000536974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000538714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000539256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
NM_001143821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
NM_001256470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
NM_001256787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
NM_019012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000299275.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000424268.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000429027.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000536974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000538714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
ENST00000539256.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
NM_001143821.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
NM_001256470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
NM_001256787.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
NM_019012.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200149072.hg.1 |
TC1200007010.hg.1 |
Clariom D TC |