Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143821.2 | NP_001137293.2 | 21 - 22 | 2803 | 123 | |
NM_001256470.1 | NP_001243399.1 | 25 - 26 | 3127 | 123 | ||
NM_001256787.1 | NP_001243716.1 | 19 - 20 | 2683 | 123 | ||
NM_019012.5 | NP_061885.2 | 19 - 20 | 2629 | 123 | ||
XM_005253400.1 | XP_005253457.1 | 20 - 21 | 2778 | 123 | ||
XM_006719091.1 | XP_006719154.1 | 21 - 22 | 2832 | 123 | ||
XM_006719093.1 | XP_006719156.1 | 19 - 20 | 2694 | 123 | ||
XM_006719096.1 | XP_006719159.1 | 24 - 25 | 3069 | 123 | ||
XM_011520714.1 | XP_011519016.1 | 23 - 24 | 3033 | 123 | ||
XM_011520715.1 | XP_011519017.1 | 22 - 23 | 2952 | 123 | ||
XM_011520716.1 | XP_011519018.1 | 23 - 24 | 2880 | 123 | ||
XM_011520717.1 | XP_011519019.1 | 21 - 22 | 2763 | 123 | ||
XM_017019495.1 | XP_016874984.1 | 24 - 25 | 3054 | 123 | ||
XM_017019496.1 | XP_016874985.1 | 23 - 24 | 2865 | 123 | ||
XM_017019497.1 | XP_016874986.1 | 22 - 23 | 2883 | 123 | ||
XM_017019498.1 | XP_016874987.1 | 18 - 19 | 2461 | 123 | ||
GenBank mRNA | AB051473.2 | - | 21 - 22 | 2699 | 123 | |
AB642244.1 | - | 25 - 26 | 3093 | 123 | ||
AF302150.1 | - | 19 - 20 | 2527 | 123 | ||
AK023127.1 | - | 7 - 8 | 650 | 123 | ||
AK056054.1 | - | 13 - 14 | 1644 | 123 | ||
AK294739.1 | - | 20 - 21 | 2684 | 123 | ||
AK295151.1 | - | 25 - 26 | 3036 | 123 | ||
AK296277.1 | - | 18 - 19 | 2595 | 123 | ||
AK297391.1 | - | 17 - 18 | 2270 | 123 | ||
AL834259.1 | - | 6 - 7 | 676 | 123 | ||
BC004164.2 | - | 10 - 11 | 1314 | 123 | ||
BC013133.2 | - | 8 - 9 | 761 | 123 | ||
BC127091.1 | - | 18 - 19 | 2454 | 40123820 | 123 | |
BC127092.1 | - | 19 - 20 | 2527 | 40123822 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |