Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000578001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000578001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000578001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000578001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000583487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000583487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000583487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
ENST00000583487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004111.hg.1 |
ENST00000254928.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004111.hg.1 |
ENST00000461894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004111.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17004111.hg.1 |
TC17000332.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700152741.hg.1 |
TC1700007399.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000254928.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000412138.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000461894.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000471992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000577942.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000578001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000578001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000578001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000578001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000580917.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000583487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000583487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000583487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
ENST00000583487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007399.hg.1 |
NM_005702.3 |
Refseq |