Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009148.hg.1 |
TC04000631.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009148.hg.1 |
ENST00000274008.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009148.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04009148.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008620.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159274.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159274.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159274.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159274.hg.1 |
ENST00000274008.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159274.hg.1 |
ENST00000422835.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400159274.hg.1 |
NM_145207.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400159274.hg.1 |
TC0400008620.hg.1 |
Clariom D TC |