Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000449027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000449027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
ENST00000449027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024654.hg.1 |
TC06001988.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024654.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024654.hg.1 |
ENST00000369033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024654.hg.1 |
ENST00000369037.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024654.hg.1 |
ENST00000453874.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024654.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000449027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000449027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
ENST00000449027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012752.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188001.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188001.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188001.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188001.hg.1 |
ENST00000369031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188001.hg.1 |
ENST00000369037.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188001.hg.1 |
NM_020381.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188001.hg.1 |
TC0600012752.hg.1 |
Clariom D TC |