Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010893.2 | NP_001010893.1 | 1 | 226 | 121 | |
GenBank mRNA | AK095808.1 | - | 1 | 1431 | 121 | |
AY825924.1 | - | 1 | 226 | 121 | ||
BC136625.1 | - | 1 | 306 | 9020615 | 121 | |
BC136631.1 | - | 1 | 306 | 9020621 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06252696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535498 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16110789 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06252695 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535532 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06252716 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16110795 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535521 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535506 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535509 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535503 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535508 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535515 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06252718 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535529 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535520 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16110791 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535530 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535528 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535510 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535511 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535524 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535512 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535527 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06252708 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06252715 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26535518 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 139528 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3416331 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0847 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0848 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 40813 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P32296465 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3416331 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3416331 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535519 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535508 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535511 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535509 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535520 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16110789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535527 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06252708 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06252696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535512 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535498 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06252715 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535510 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535515 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06252695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535518 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535518 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06252708 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535527 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06252695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535511 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535510 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06252696 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16110789 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535512 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535498 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535519 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535508 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535509 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06252715 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535515 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26535520 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P01954681 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06252693 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P06252697 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01954681 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 34564 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 6750 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38465804 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32296465 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P736843 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252709 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252704 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252702 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252701 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252706 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252655 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252712 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252699 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252693 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252665 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252654 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252671 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252722 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252720 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252715 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252669 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252661 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16110797 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252695 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252656 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252658 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252652 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252718 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252708 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06252711 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P32296465 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001010893_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151560 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8151559 | - | - |
Affymetrix Human Genome U219 Array | 11738298_at | - | - |
Affymetrix Human Genome U219 Array | 11738298_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0800010945.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010945.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010945.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010945.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010945.hg.1 | NM_001010893.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08017668.hg.1 | TC08001369.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08017668.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08017668.hg.1 | NM_001010893.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174369.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174369.hg.1 | NM_001010893.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174369.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174369.hg.1 | NM_001010893.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800174369.hg.1 | TC0800010945.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800174370.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174370.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800174370.hg.1 | TC0800010945.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800010945.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010945.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010945.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010945.hg.1 | ENST00000518568.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010945.hg.1 | NM_001010893.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035910 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035910 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 233094 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1737323 | AY825924.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1737323 | BC136625.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1737323 | BC136631.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_13121-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 840332 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_13121 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1737323 | AK095808.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1737323 | AY825924.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1737323 | BC136625.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1737323 | BC136631.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1737323 | NM_001010893.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1737323 | AK095808.1 | human_transcriptome |