Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002555.hg.1 |
TC02000183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002555.hg.1 |
ENST00000264716.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002555.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002556.hg.1 |
TC02000183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002556.hg.1 |
ENST00000264716.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002556.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002559.hg.1 |
TC02000183.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002559.hg.1 |
ENST00000264716.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002559.hg.1 |
ENST00000379619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002559.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002559.hg.1 |
ENST00000545753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002559.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148822.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148822.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148822.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148822.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148822.hg.1 |
TC0200007096.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200148824.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148824.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148824.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148824.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148824.hg.1 |
TC0200007096.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007096.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148823.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148823.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148823.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148823.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148823.hg.1 |
TC0200007096.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
ENST00000264716.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
ENST00000379619.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
ENST00000436647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
ENST00000460736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
NM_005253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148826.hg.1 |
TC0200007096.hg.1 |
Clariom D TC |