Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019498.hg.1 |
TC02001273.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019498.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176187.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176187.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176187.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176187.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176187.hg.1 |
TC0200010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176190.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176190.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176190.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176190.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176190.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176190.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176190.hg.1 |
TC0200010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176192.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176192.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176192.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176192.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176192.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176192.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176192.hg.1 |
TC0200010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010734.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176189.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176189.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176189.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176189.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176189.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176189.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176189.hg.1 |
TC0200010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176188.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176188.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176188.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176188.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176188.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176188.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176188.hg.1 |
TC0200010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176186.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176186.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176186.hg.1 |
TC0200010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176185.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176185.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176185.hg.1 |
TC0200010734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200176191.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176191.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176191.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176191.hg.1 |
ENST00000562038.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176191.hg.1 |
ENST00000597904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176191.hg.1 |
NR_037701.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176191.hg.1 |
TC0200010734.hg.1 |
Clariom D TC |