Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000339764.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000535730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000537753.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000542350.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
ENST00000543947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008400.hg.1 |
NM_153614.3 |
Refseq |