Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002387.hg.1 |
TC04000152.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002387.hg.1 |
ENST00000237380.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04002387.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000499786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000499786.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000506409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000506409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000506409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000499786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000499786.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000506409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000506409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000506409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006990.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
ENST00000237380.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
ENST00000503945.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
ENST00000612711.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
NM_025205.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400147966.hg.1 |
TC0400006990.hg.1 |
Clariom D TC |