Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019173.hg.1 |
TC04001370.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019173.hg.1 |
ENST00000237612.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019173.hg.1 |
ENST00000515655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019173.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177131.hg.1 |
TC0400011279.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000237612.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000503830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000505480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
ENST00000515655.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_001257386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011279.hg.1 |
NM_004827.2 |
Refseq |