Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000526823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000526823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000526823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000526823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000530950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000530950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000530950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000530950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000532895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000532895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
ENST00000532895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
NM_021571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
NM_021571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
NM_021571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012170.hg.1 |
NM_021571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000526823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000526823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000526823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000526823.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000530950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000530950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000530950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000530950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000532895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000532895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
ENST00000532895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
NM_021571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
NM_021571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
NM_021571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012170.hg.1 |
NM_021571.3 |
Refseq |