Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000512233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000512233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
ENST00000512233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012612.hg.1 |
TC04000925.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012612.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04012612.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000507581.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000512233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000512233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
ENST00000512233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009592.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165748.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165748.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165748.hg.1 |
ENST00000332517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400165748.hg.1 |
NM_178556.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400165748.hg.1 |
TC0400009592.hg.1 |
Clariom D TC |