Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178140.3 | NP_835260.2 | 15 - 16 | 2919 | 110 | |
XM_005248269.4 | XP_005248326.1 | 15 - 16 | 2941 | 110 | ||
XM_005248271.1 | XP_005248328.1 | 14 - 15 | 2293 | 110 | ||
XM_005248272.3 | XP_005248329.1 | 14 - 15 | 2295 | 110 | ||
XM_006714460.2 | XP_006714523.1 | 14 - 15 | 2092 | 110 | ||
XM_011513992.2 | XP_011512294.1 | 16 - 17 | 3303 | 110 | ||
XM_011513993.2 | XP_011512295.1 | 15 - 16 | 3221 | 110 | ||
XM_011513994.2 | XP_011512296.1 | 16 - 17 | 3304 | 110 | ||
XM_011513995.2 | XP_011512297.1 | 16 - 17 | 3302 | 110 | ||
XM_011513996.2 | XP_011512298.1 | 14 - 15 | 3027 | 110 | ||
XM_017009245.1 | XP_016864734.1 | 12 - 13 | 2017 | 110 | ||
XM_017009246.1 | XP_016864735.1 | 16 - 17 | 3219 | 110 | ||
GenBank mRNA | AB002298.2 | - | 15 - 16 | 2919 | 110 | |
AK091490.1 | - | 3 - 4 | 313 | 110 | ||
AK304804.1 | - | 14 - 15 | 2288 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Homo sapiens Probe spans exons |