Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059399.hg.1 |
TC01003896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059399.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059399.hg.1 |
ENST00000366818.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059399.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059398.hg.1 |
TC01003896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059398.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059398.hg.1 |
ENST00000366817.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059398.hg.1 |
ENST00000366818.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059398.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059400.hg.1 |
TC01003896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059400.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059400.hg.1 |
ENST00000366818.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059400.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232534.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232534.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232534.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232534.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232534.hg.1 |
TC0100017505.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
ENST00000272091.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017505.hg.1 |
NM_152608.3 |
Refseq |