Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013973.hg.1 |
TC03000765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013973.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013973.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013974.hg.1 |
TC03000765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013974.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013974.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013972.hg.1 |
TC03000765.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013972.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013972.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164663.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164663.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164663.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164663.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164663.hg.1 |
TC0300009012.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300164666.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164666.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164666.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164666.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164666.hg.1 |
TC0300009012.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300164665.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164665.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164665.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300164665.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300164665.hg.1 |
TC0300009012.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
ENST00000264952.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009012.hg.1 |
NM_139209.2 |
Refseq |