Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007415.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153061.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153061.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153061.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153061.hg.1 |
ENST00000316436.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300153061.hg.1 |
NM_001304385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153061.hg.1 |
NM_153215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300153061.hg.1 |
TC0300007415.hg.1 |
Clariom D TC |