Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144939.hg.1 |
TC1600006486.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000219551.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000352681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000450775.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
ENST00000561556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006486.hg.1 |
NM_001278721.1 |
Refseq |