Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026187.hg.1 |
TC01001687.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026187.hg.1 |
ENST00000367222.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026187.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026190.hg.1 |
TC01001687.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026190.hg.1 |
ENST00000367222.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026190.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182683.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182683.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182683.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182683.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182683.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182683.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182683.hg.1 |
TC0100011263.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100182679.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182679.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182679.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182679.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182679.hg.1 |
TC0100011263.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011263.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182682.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182682.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182682.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182682.hg.1 |
ENST00000367222.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182682.hg.1 |
ENST00000448911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182682.hg.1 |
NM_014359.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182682.hg.1 |
TC0100011263.hg.1 |
Clariom D TC |