Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019800.hg.1 |
ENST00000380103.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019800.hg.1 |
ENST00000398453.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019800.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019800.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12019800.hg.1 |
TC12001506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181575.hg.1 |
TC1200010702.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000604188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010702.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000380103.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000398453.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000603798.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000603864.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
ENST00000605627.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
NM_001031628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
NM_001033873.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200181574.hg.1 |
TC1200010702.hg.1 |
Clariom D TC |