Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010606.hg.1 |
TC09000820.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010606.hg.1 |
ENST00000371691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010606.hg.1 |
ENST00000371692.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010606.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010605.hg.1 |
TC09000820.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010605.hg.1 |
ENST00000371691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010605.hg.1 |
ENST00000371692.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09010605.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162912.hg.1 |
TC0900009231.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009231.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
ENST00000371691.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
ENST00000371692.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
ENST00000617017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
NM_152421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162913.hg.1 |
TC0900009231.hg.1 |
Clariom D TC |