Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000473695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000473695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000473695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000473695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000475023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000475023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000475023.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016480.hg.1 |
TC09001324.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016480.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016480.hg.1 |
ENST00000375731.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016480.hg.1 |
ENST00000422391.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016480.hg.1 |
ENST00000478465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016480.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
ENST00000475023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
ENST00000475023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173194.hg.1 |
TC0900010762.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000303617.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000375731.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000473695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000473695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000473695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000473695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000475023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000475023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000475023.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
ENST00000478465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001306190.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010762.hg.1 |
NM_001698.2 |
Refseq |