Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004398.hg.1 |
ENST00000361320.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004398.hg.1 |
ENST00000373722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004398.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004398.hg.1 |
TC10000396.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004399.hg.1 |
ENST00000361320.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004399.hg.1 |
ENST00000373722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004399.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004399.hg.1 |
TC10000396.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004397.hg.1 |
ENST00000361320.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004397.hg.1 |
ENST00000373722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004397.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004397.hg.1 |
TC10000396.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000472278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000472278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000472278.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004400.hg.1 |
ENST00000361320.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004400.hg.1 |
ENST00000373722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004400.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10004400.hg.1 |
TC10000396.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000152974.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152974.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152974.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152974.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152974.hg.1 |
TC1000007833.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000152976.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152976.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152976.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152976.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152976.hg.1 |
TC1000007833.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000152978.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152978.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152978.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152978.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152978.hg.1 |
TC1000007833.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000152975.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152975.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152975.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000152975.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000152975.hg.1 |
TC1000007833.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000361320.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000472278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000472278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000472278.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
ENST00000485868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NM_178011.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007833.hg.1 |
NR_111909.1 |
Refseq |