Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023683.hg.1 |
TC02001471.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023683.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02023683.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200182107.hg.1 |
TC0200011376.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000307503.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000470255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000472436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
ENST00000476698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011376.hg.1 |
NM_000030.2 |
Refseq |