Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000261658.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000562545.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563313.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000563971.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000564078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000564078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000564078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000564078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000564078.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000565478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000565478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000565478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000565478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000566710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000570219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000570219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000570219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
ENST00000570219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006995.hg.1 |
NM_016561.2 |
Refseq |