Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000300469.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000300469.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000300469.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008843.hg.1 |
ENST00000588017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008843.hg.1 |
ENST00000588017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006590.hg.1 |
ENST00000545761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003091.hg.1 |
ENST00000357156.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003091.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003091.hg.1 |
ENST00000417166.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003091.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003091.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003091.hg.1 |
TC20000242.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000149348.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149348.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149348.hg.1 |
TC2000007191.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149348.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149348.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008843.hg.1 |
ENST00000588017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008843.hg.1 |
ENST00000588017.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006590.hg.1 |
ENST00000545761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
TC2000007191.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149359.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000300469.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000300469.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000300469.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000357156.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000374846.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
ENST00000480759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281727.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_001281729.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_014183.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NM_177953.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007191.hg.1 |
NR_104032.1 |
Refseq |