Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025787.hg.1 |
TC01001665.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025787.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025787.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025787.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025787.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025782.hg.1 |
TC01001665.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025782.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025782.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025782.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025782.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182086.hg.1 |
TC0100011197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182080.hg.1 |
TC0100011197.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000362011.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000464117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
ENST00000481699.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NM_198149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011197.hg.1 |
NR_030775.1 |
Refseq |