Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012295.hg.1 |
ENST00000173785.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012295.hg.1 |
ENST00000497571.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012295.hg.1 |
NM_001160124.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012295.hg.1 |
NM_001300.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012295.hg.1 |
NR_027653.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012295.hg.1 |
TC10000979.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
ENST00000173785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
ENST00000461124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
ENST00000492125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
ENST00000497571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
NM_001160124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
NM_001300.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
NR_027653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
ENST00000173785.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
ENST00000461124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
ENST00000492125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
ENST00000497571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
NM_001160124.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
NM_001300.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
NR_027653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000165685.hg.1 |
TC1000009612.hg.1 |
Clariom D TC |