Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027979.hg.1 |
TC01001790.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027979.hg.1 |
ENST00000366957.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027979.hg.1 |
ENST00000415093.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027979.hg.1 |
ENST00000491455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027979.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000416415.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000484459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011573.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
ENST00000366957.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
ENST00000460580.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
ENST00000471645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
ENST00000491455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
NM_020197.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185236.hg.1 |
TC0100011573.hg.1 |
Clariom D TC |