Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300743.1 | NP_001287672.1 | 5 - 6 | 750 | 104 | |
NM_020314.5 | NP_064710.4 | 5 - 6 | 750 | 104 | ||
XM_005255435.3 | XP_005255492.1 | 5 - 6 | 702 | 104 | ||
XM_006721065.3 | XP_006721128.1 | 5 - 6 | 702 | 104 | ||
XM_011545896.2 | XP_011544198.1 | 4 - 5 | 380 | 104 | ||
GenBank mRNA | AK095078.1 | - | 5 - 6 | 447 | 104 | |
AK290286.1 | - | 5 - 6 | 447 | 104 | ||
AL832159.1 | - | 5 - 6 | 446 | 104 | ||
AL833428.1 | - | 5 - 6 | 439 | 104 | ||
BC014900.2 | - | 4 - 5 | 418 | 3909068 | 104 | |
BC050464.1 | - | 5 - 6 | 490 | 6061139 | 104 | |
BC058845.1 | - | 5 - 6 | 449 | 6452778 | 104 | |
BX537867.1 | - | 5 - 6 | 462 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |