Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005918.hg.1 |
TC06000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005918.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005918.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005922.hg.1 |
TC06000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005922.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005922.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005922.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005920.hg.1 |
TC06000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005920.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005920.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005920.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005923.hg.1 |
TC06000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005923.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005923.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06005923.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154612.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154612.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154612.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154612.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154612.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154612.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154612.hg.1 |
TC0600007862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154613.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154613.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154613.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154613.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154613.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154613.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154613.hg.1 |
TC0600007862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000468243.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
ENST00000479509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007862.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154614.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154614.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154614.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154614.hg.1 |
ENST00000373509.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600154614.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154614.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154614.hg.1 |
TC0600007862.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600154610.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154610.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154610.hg.1 |
NM_001243186.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154610.hg.1 |
NM_002648.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600154610.hg.1 |
TC0600007862.hg.1 |
Clariom D TC |