Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 266 | TMEM266 |
C15orf27, HVRP1 |
-2.0kb | Anti-sense | Chr. 15 - 76059929 - 76204963 on NCBI Build GRCh38 |
Downstream | electron transfer flavoprotein alpha subunit | ETFA |
EMA, GA2, MADD |
6.0kb | Sense | Chr. 15 - 76216228 - 76311469 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |