Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007141.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148894.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148894.hg.1 |
NM_182658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148894.hg.1 |
TC2000007141.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148894.hg.1 |
ENST00000375494.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148894.hg.1 |
NM_182658.2 |
Refseq |