Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001879.hg.1 |
ENST00000267889.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001879.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001879.hg.1 |
TC15000281.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000558623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000558623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500181793.hg.1 |
ENST00000559721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500181793.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500181793.hg.1 |
ENST00000559721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500181793.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500181793.hg.1 |
TC1500010717.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000558623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000558623.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
ENST00000559721.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010717.hg.1 |
NM_033510.2 |
Refseq |