Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007208.hg.1 |
TC01000466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007208.hg.1 |
ENST00000207457.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007208.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007790.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
ENST00000207457.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
ENST00000469024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
ENST00000473120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
NM_014466.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154887.hg.1 |
TC0100007790.hg.1 |
Clariom D TC |